Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRTDAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024642.1 | NP_001019813.1 | 4 - 5 | 227 | 79 | |
GenBank mRNA | AY999063.1 | - | 4 - 5 | 227 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Canis familiaris Probe spans exons |