Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTBDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024961.1 | NP_001020132.1 | 3 - 4 | 327 | 66 | |
XM_005632322.2 | XP_005632379.1 | 4 - 5 | 612 | 66 | ||
XM_014121734.2 | XP_013977209.1 | 5 - 6 | 2783 | 66 | ||
XM_022406232.1 | XP_022261940.1 | 5 - 6 | 2413 | 66 | ||
XM_022406233.1 | XP_022261941.1 | 3 - 4 | 1185 | 66 | ||
GenBank mRNA | DQ058011.1 | - | 3 - 4 | 327 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |