Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC45A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037947.1 | NP_001033036.1 | 1 - 2 | 456 | 68 | |
XM_005618882.3 | XP_005618939.1 | 1 - 2 | 1832 | 68 | ||
GenBank mRNA | DQ302162.1 | - | 1 - 2 | 456 | 68 | |
KM222821.1 | - | 1 - 2 | 424 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |