Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNLIPRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003319.2 | NP_001003319.2 | 12 - 13 | 1369 | 60 | |
GenBank mRNA | DN329403.1 | - | 2 - 3 | 206 | 60 | |
M35302.1 | - | 12 - 13 | 1375 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Canis familiaris Probe spans exons |