Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNLIPRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003319.2 | NP_001003319.2 | 9 - 10 | 957 | 75 | |
GenBank mRNA | M35302.1 | - | 9 - 10 | 963 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Canis familiaris Probe spans exons |