Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FXYD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195142.1 | NP_001182071.1 | 2 - 3 | 70 | ||
NM_001195143.1 | NP_001182072.1 | 2 - 3 | 70 | |||
XM_005615754.3 | XP_005615811.2 | 2 - 3 | 70 | |||
XM_005615755.3 | XP_005615812.2 | 2 - 3 | 70 | |||
XM_014114177.2 | XP_013969652.2 | 2 - 3 | 70 | |||
XM_014114188.2 | XP_013969663.1 | 2 - 3 | 70 | |||
GenBank mRNA | M63934.1 | - | 2 - 3 | 164 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Canis familiaris Probe spans exons |