Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2B6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006652.1 | NP_001006653.1 | 4 - 5 | 653 | 74 | |
GenBank mRNA | HG965162.1 | - | 4 - 5 | 645 | 74 | |
HG965163.1 | - | 4 - 5 | 645 | 74 | ||
HG974426.1 | - | 4 - 5 | 645 | 74 | ||
M92447.1 | - | 4 - 5 | 653 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |