Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003004.1 | NP_001003004.1 | 2 - 3 | 431 | 107 | |
XM_005636669.3 | XP_005636726.1 | 2 - 3 | 1142 | 107 | ||
GenBank mRNA | DQ672621.1 | - | 2 - 3 | 432 | 107 | |
EU285563.1 | - | 2 - 3 | 432 | 107 | ||
U52916.1 | - | 2 - 3 | 431 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |