Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003231.2 | NP_001003231.2 | 7 - 8 | 684 | 141 | |
GenBank mRNA | U77716.2 | - | 686 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Probe spans exons |