Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003231.2 | NP_001003231.2 | 8 - 9 | 817 | 88 | |
GenBank mRNA | U77716.2 | - | 819 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Canis familiaris Probe spans exons |