Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003276.3 | NP_001003276.1 | 3 - 4 | 542 | 68 | |
GenBank mRNA | CO591375.1 | - | 2 - 3 | 475 | 68 | |
X56388.1 | - | 443 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Canis familiaris Probe spans exons |