Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003276.3 | NP_001003276.1 | 4 - 5 | 624 | 103 | |
GenBank mRNA | CO591375.1 | - | 1 - 2 | 393 | 103 | |
X56388.1 | - | 525 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |