Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003274.2 | NP_001003274.1 | 1 | 37 | 85 | |
XM_005621021.1 | XP_005621078.1 | 85 | ||||
GenBank mRNA | X56390.1 | - | 37 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |