Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003265.2 | NP_001003265.1 | 619 | 95 | ||
GenBank mRNA | X72933.1 | - | 619 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris |