Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022421726.1 | XP_022277434.1 | 4 - 5 | 487 | 118 | |
GenBank mRNA | Z22818.1 | - | 319 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |