Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005616460.3 | XP_005616517.2 | 4 | 553 | 98 | |
XM_022425020.1 | XP_022280728.1 | 4 | 552 | 98 | ||
XM_533644.6 | XP_533644.1 | 4 | 566 | 98 | ||
XM_860950.4 | XP_866043.1 | 4 | 580 | 98 | ||
GenBank mRNA | AY926587.1 | - | 1 | 49 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |