Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003266.2 | NP_001003266.1 | 4 | 667 | 155 | |
GenBank mRNA | X56994.1 | - | 369 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |