Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FABP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005617665.3 | XP_005617722.1 | 3 - 4 | 431 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris Probe spans exons |