Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005631426.3 | XP_005631483.1 | 1 - 2 | 172 | 80 | |
XM_022405615.1 | XP_022261323.1 | 1 - 2 | 172 | 80 | ||
XM_022405618.1 | XP_022261326.1 | 1 - 2 | 180 | 80 | ||
XM_533216.6 | XP_533216.1 | 1 - 2 | 170 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Canis familiaris Probe spans exons |