Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHOC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005637720.3 | XP_005637777.1 | 7 - 8 | 1855 | 77 | |
XM_022411763.1 | XP_022267471.1 | 6 - 7 | 1611 | 77 | ||
XM_535013.6 | XP_535013.2 | 7 - 8 | 1749 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |