Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014114701.2 | XP_013970176.1 | 1 - 2 | 157 | 79 | |
XM_022420271.1 | XP_022275979.1 | 1 - 2 | 157 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |