Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UQCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005634970.3 | XP_005635027.1 | 2 - 3 | 264 | 95 | |
XM_005634971.3 | XP_005635028.1 | 2 - 3 | 264 | 95 | ||
XM_014106999.1 | XP_013962474.1 | 2 - 3 | 264 | 95 | ||
XM_014107002.2 | XP_013962477.1 | 2 - 3 | 264 | 95 | ||
XM_846758.5 | XP_851851.1 | 2 - 3 | 264 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |