Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UQCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005634970.3 | XP_005635027.1 | 6 - 7 | 590 | 121 | |
XM_005634971.3 | XP_005635028.1 | 6 - 7 | 590 | 121 | ||
XM_014106999.1 | XP_013962474.1 | 6 - 7 | 590 | 121 | ||
XM_014107000.2 | XP_013962475.1 | 5 - 6 | 494 | 121 | ||
XM_014107002.2 | XP_013962477.1 | 6 - 7 | 590 | 121 | ||
XM_846758.5 | XP_851851.1 | 6 - 7 | 590 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |