Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313778.1 | NP_001300707.1 | 2 - 3 | 221 | 75 | |
XM_857783.4 | XP_862876.1 | 2 - 3 | 319 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Canis familiaris Probe spans exons |