Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005636909.3 | XP_005636966.1 | 3 - 4 | 391 | 70 | |
XM_005636910.3 | XP_005636967.1 | 4 - 5 | 538 | 70 | ||
XM_005636911.3 | XP_005636968.1 | 4 - 5 | 534 | 70 | ||
XM_005636913.3 | XP_005636970.1 | 3 - 4 | 358 | 70 | ||
XM_022410952.1 | XP_022266660.1 | 4 - 5 | 576 | 70 | ||
XM_022410953.1 | XP_022266661.1 | 3 - 4 | 445 | 70 | ||
XM_022410954.1 | XP_022266662.1 | 2 - 3 | 184 | 70 | ||
XM_543713.5 | XP_543713.2 | 3 - 4 | 359 | 70 | ||
XM_858305.5 | XP_863398.1 | 4 - 5 | 460 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |