Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_534131.6 | XP_534131.1 | 4 - 5 | 337 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |