Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OGFOD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014107935.2 | XP_013963410.1 | 177 | 115 | ||
XM_534653.6 | XP_534653.3 | 176 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris |