Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC12A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_534891.6 | XP_534891.2 | 4 - 5 | 720 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris Probe spans exons |