Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPSB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005637212.3 | XP_005637269.1 | 2 - 3 | 879 | 57 | |
XM_005637213.3 | XP_005637270.1 | 2 - 3 | 884 | 57 | ||
XM_005637214.3 | XP_005637271.1 | 2 - 3 | 819 | 57 | ||
XM_022410961.1 | XP_022266669.1 | 2 - 3 | 771 | 57 | ||
XM_022410962.1 | XP_022266670.1 | 2 - 3 | 995 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Canis familiaris Probe spans exons |