Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAUS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022412532.1 | XP_022268240.1 | 4 - 5 | 427 | 79 | |
XM_022412533.1 | XP_022268241.1 | 4 - 5 | 723 | 79 | ||
XM_022412534.1 | XP_022268242.1 | 4 - 5 | 621 | 79 | ||
XM_535444.6 | XP_535444.1 | 4 - 5 | 427 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Canis familiaris Probe spans exons |