Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYYR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_535572.6 | XP_535572.1 | 2 - 3 | 263 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |