Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022413390.1 | XP_022269098.1 | 10 - 11 | 1332 | 124 | |
XM_535620.6 | XP_535620.1 | 8 - 9 | 1529 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris Probe spans exons |