Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDOC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286945.1 | NP_001273874.1 | 2 - 3 | 251 | 86 | |
XM_022422577.1 | XP_022278285.1 | 3 - 4 | 370 | 86 | ||
XM_022422578.1 | XP_022278286.1 | 3 - 4 | 364 | 86 | ||
GenBank mRNA | GR887893.1 | - | 2 - 3 | 250 | 86 | |
GR893395.1 | - | 2 - 3 | 251 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |