Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUZD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_544054.6 | XP_544054.2 | 8 - 9 | 1719 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris Probe spans exons |