Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDOC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286945.1 | NP_001273874.1 | 4 - 5 | 517 | 79 | |
XM_022422577.1 | XP_022278285.1 | 5 - 6 | 636 | 79 | ||
XM_022422578.1 | XP_022278286.1 | 5 - 6 | 630 | 79 | ||
GenBank mRNA | GR887893.1 | - | 4 - 5 | 516 | 79 | |
GR893395.1 | - | 4 - 5 | 517 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Canis familiaris Probe spans exons |