Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLAGL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005615474.3 | XP_005615531.1 | 338 | 63 | ||
XM_014112545.2 | XP_013968020.1 | 338 | 63 | |||
XM_014112555.2 | XP_013968030.1 | 376 | 63 | |||
XM_014112558.2 | XP_013968033.1 | 408 | 63 | |||
XM_014112566.2 | XP_013968041.1 | 370 | 63 | |||
XM_014112572.2 | XP_013968047.1 | 352 | 63 | |||
XM_022417720.1 | XP_022273428.1 | 304 | 63 | |||
XM_022417724.1 | XP_022273432.1 | 279 | 63 | |||
XM_022417725.1 | XP_022273433.1 | 202 | 63 | |||
XM_022417731.1 | XP_022273439.1 | 336 | 63 | |||
XM_022417736.1 | XP_022273444.1 | 318 | 63 | |||
XM_022417744.1 | XP_022273452.1 | 338 | 63 | |||
XM_022417747.1 | XP_022273455.1 | 338 | 63 | |||
XM_022417748.1 | XP_022273456.1 | 338 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris |