Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGSN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048103.1 | NP_001041568.1 | 2 - 3 | 187 | 90 | |
XM_014118049.2 | XP_013973524.1 | 2 - 3 | 277 | 90 | ||
GenBank mRNA | DQ415892.1 | - | 2 - 3 | 187 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |