Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFX8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022423975.1 | XP_022279683.1 | 3 - 4 | 273 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |