Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047981.1 | NP_001041446.1 | 6 - 7 | 513 | 114 | |
XM_014114134.2 | XP_013969609.1 | 5 - 6 | 524 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |