Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_531866.6 | XP_531866.2 | 562 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris |