Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TREM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_532133.6 | XP_532133.2 | 1 - 2 | 175 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |