Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014117997.2 | XP_013973472.1 | 3 - 4 | 347 | 81 | |
XM_533554.6 | XP_533554.1 | 4 - 5 | 408 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Canis familiaris Probe spans exons |