Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGCG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014276.2 | NP_001014298.1 | 387 | 65 | ||
XM_005635383.3 | XP_005635440.1 | 589 | 65 | |||
XM_005635384.3 | XP_005635441.1 | 553 | 65 | |||
XM_022409561.1 | XP_022265269.1 | 641 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris |