Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYROBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197117.1 | NP_001184046.1 | 1 - 2 | 148 | 87 | |
XM_005615749.3 | XP_005615806.1 | 1 - 2 | 166 | 87 | ||
GenBank mRNA | CO655102.1 | - | 4 - 5 | 443 | 87 | |
CO704168.1 | - | 4 - 5 | 444 | 87 | ||
DN439202.1 | - | 1 - 2 | 140 | 87 | ||
FJ687210.1 | - | 1 - 2 | 65 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Canis familiaris Inventoried Probe spans exons |