Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCFD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005626084.3 | XP_005626141.2 | 2 - 3 | 396 | 67 | |
XM_531811.4 | XP_531811.1 | 2 - 3 | 251 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |