Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLIPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005625646.2 | XP_005625703.1 | 889 | 104 | ||
XM_531682.6 | XP_531682.1 | 1090 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris |