Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYRK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003434949.4 | XP_003434997.2 | 249 | 94 | ||
XM_014115059.2 | XP_013970534.1 | 261 | 94 | |||
XM_022420843.1 | XP_022276551.1 | 231 | 94 | |||
XM_022420844.1 | XP_022276552.1 | 147 | 94 |