Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014111886.2 | XP_013967361.1 | 3 - 4 | 215 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Probe spans exons |