Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VMA21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252312.1 | NP_001239241.1 | 1 - 2 | 140 | 72 | |
GenBank mRNA | BM538662.1 | - | 1 - 2 | 45 | 72 | |
DN384812.1 | - | 1 - 2 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |