Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNIH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005622974.2 | XP_005623031.2 | 2 - 3 | 267 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris Probe spans exons |